missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate, pure, 10 g/l solution in sulfuric acid, Thermo Scientific Chemicals

£96.88 - £241.20

Specifications

Chemical Name or Material Silver sulfate
Color Colorless
Name Note 10g/L Solution in Sulfuric Acid
CAS 10294-26-5, 7664-93-9
Molecular Formula Ag2H2O4S
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10237460
View Documents Promotion Details
Thermo Scientific Acros
279480010
1 L Glass bottle
£96.88
1 L
Please to purchase this item. Need a web account? Register with us today!
10247460
View Documents
Thermo Scientific Acros
279480025
2.5 L Glass bottle
£241.20
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver sulfate
10g/L Solution in Sulfuric Acid
Ag2H2O4S
MFCD00003407
1.8400g/mL
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
[Ag+].[Ag+].OS(O)(=O)=O
313.81
311.79
Glass bottle
Liquid
Colorless
10294-26-5, 7664-93-9
Ag2SO4
01,1015; 03,254; 04,435
15, 8668
YPNVIBVEFVRZPJ-UHFFFAOYSA-N
disilver(1+) sulfuric acid
159865
Pure
1.84
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 233-653-7

missing translation for 'tsca' : TSCA

Product Identifier
  • Silver sulfate, 10 g/l solution in sulfuric acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only