Learn More
Silver sulfide, Premion™, 99.995% (metals basis), Thermo Scientific Chemicals
£86.60 - £307.00
Chemical Identifiers
CAS | 21548-73-2 |
---|---|
Molecular Formula | Ag2S |
Molecular Weight (g/mol) | 247.80 |
MDL Number | MFCD00003406 |
InChI Key | XUARKZBEFFVFRG-UHFFFAOYSA-N |
IUPAC Name | disilver(1+) sulfanediide |
SMILES | [S--].[Ag+].[Ag+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356478
|
Thermo Scientific Alfa Aesar
012113.06 |
5 g |
£86.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346478
|
Thermo Scientific Alfa Aesar
012113.14 |
25 g |
£307.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds use as a photosensitizer in photography. Blackening the silver for jewelleries and disinfection/sterilization of reservoirs. Anti-bacterial and anti-microbial agents. Laboratory reagents. A mixed ionic-electronic conductor. In IR detectors, photoconductors, photovoltaic cells, electrochemical storage cells, etc. Black colorant in ceramic glazes and for niello metal work. Autometallography in the detection of catalytic tissue metals and colloidal gold particles bound to macromolecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
21548-73-2 | |
247.80 | |
XUARKZBEFFVFRG-UHFFFAOYSA-N | |
[S--].[Ag+].[Ag+] |
Ag2S | |
MFCD00003406 | |
disilver(1+) sulfanediide |
Specifications
825°C | |
Gray to Black | |
99.998% (metals basis) | |
MFCD00003406 | |
XUARKZBEFFVFRG-UHFFFAOYSA-N | |
disilver(1+) sulfanediide | |
247.8 | |
Light sensitive | |
7.317 |
21548-73-2 | |
Powder | |
Ag2S | |
Insoluble in most of the solvents. Insoluble in acids,alkalies and aqueous ammoniums. | |
[S--].[Ag+].[Ag+] | |
247.80 | |
Odorless | |
Dark grey to black powder/flake/crystalline/beads | |
Silver sulfide |
Safety and Handling
H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 244-438-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only