Learn More
Silver trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Description
Silver trifluoromethanesulfonate is useful in halide abstraction. It helps in activating sulphur protecting groups at the anomeric centres of carbohydrates in the presence of iodine. It is employed as a triflating agent in combination with Palladium compounds, as a catalyst for cross coupling reactions, and in several organic transformation reactions. Polyurethanes prepared from silver triflate exhibit high permeability and selectivity towards olefins in their separation from paraffin mixtures. Polyethylene oxide - silver triflate composites find application in humidity sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 356°C |
Color | White |
Physical Form | Crystalline |
Quantity | 50 g |
Assay Percent Range | 98% |
Beilstein | EM>10, |
Solubility Information | Soluble in water and also in most organic solvents. |
Formula Weight | 256.93 |
Odor | Odorless |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only