missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 1.0mm (0.04in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11351269
Click to view available options
Quantity:
1 m
25 m
5 m

Form: approx. 8.24g/m, Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored wire
Assay Percent Range 99.9% (Metals basis)
Quantity 5 m
Odor Characteristic
Form Wire
Chemical Name or Material Silver, Annealed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'