Learn More
Motic™ Simple Add-on Polarizer Filter
Illuminates the specimen while Analyzer restricts detected light. Motic™ Simple Add-on Polarizer Filter is for use with BA Series Microscopes
Brand:  Motic™ 1101000300741
Product Code. 11783197
  • £33.00 / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with BA Series Microscopes
  • Requires use with Analyzer
  • Illuminates the specimen while analyzer restricts detected light
Specifications
Specifications
Polarizing Filter
Specimen illumination
1 Ea.
Requires use with Analyzer Filter
BA Series Microscopes