Learn More
Applied Biosystems™ SimpliAmp™ Thermal Cycler + Plastics Packages
The Applied Biosystems SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow.
£5190.00
Specifications
Thermal Accuracy | ±0.25°C (35°C to 99.9°C) |
---|---|
High-throughput Compatibility | Manual Protocols |
For Use With (Equipment) | GeneAmp 9700, GeneAmp™ 2700, Veriti™ Thermal Cycler, 2720 Thermal Cycler |
Thermal Uniformity | <0.5°C (20 sec. after reaching 95°C) |
Dimensions | 18.1 x 9.5 x 8.3 in. (46 x 24 x 21 cm) |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17153179
|
Applied Biosystems™
A41192 |
Includes MicroAmp 8-Tube Strips with Attached Domed Caps, 0.2 mL (125 strips) |
£5190.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17163179
|
Applied Biosystems™
A41193 |
Includes 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (100 plates total) |
£5190.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Applied Biosystems SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow. Innovations like a responsive color touch-screen and VeriFlex temperature control technology enable simple, accurate optimization. And now you can access your SimpliAmp cycler with Thermo Fisher Connect, giving you the freedom to design and securely upload your methods from any mobile device or desktop computer.
SimpliAmp Thermal Cycler
This SimpliAmp Thermal Cycler package includes a thermal cycler and either a MicroAmp EnduraPlates with MicroAmp Clear Adhesive Films or MicroAmp 8-Tube Strips with Attached Domed Caps.
MicroAmp 8-Tube Strips with Attached Domed Caps
MicroAmp 8-Tube Strips with Attached Domed Caps feature individually attached domed caps, dual large end-tabs, and graduated 20-μL markers on each tube to help prevent cross-contamination, pipetting errors, and sample identification errors.
• Strip of eight tubes with attached domed caps that open and close independently to prevent contamination and evaporation
• Angled caps that provide efficient tube strip positioning in racks and thermal cycler blocks
• Etched A-H letter labeling on individual tubes visible when caps are open or closed
• Dual end tabs for proper labeling and easy handling
• Graduated 20-μL measuring markers on each tube for visual volume verification
MicroAmp EnduraPlates
MicroAmp EnduraPlates offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. They are a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments. They are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.
• Optimized for performance with Applied Biosystems instruments
• Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete workflow needs
MicroAmp Clear Adhesive Films
MicroAmp Clear Adhesive Films create a secure seal across an entire microplate to prevent evaporation and well-to-well contamination.
• Adhesive provides tight seal over wells to prevent leaks and evaporation.
• Easy to apply and remove
• Available as Clear (economical and sticky) or Optical (ready for real-time PCR and non-tacky)
Specifications
±0.25°C (35°C to 99.9°C) | |
GeneAmp 9700, GeneAmp™ 2700, Veriti™ Thermal Cycler, 2720 Thermal Cycler | |
18.1 x 9.5 x 8.3 in. (46 x 24 x 21 cm) | |
100/240 V | |
Thermal Cycler | |
0°C to 100°C | |
600 W | |
>1000 | |
96-well, 3-Zone VeriFlex Block |
Manual Protocols | |
<0.5°C (20 sec. after reaching 95°C) | |
96-well, 0.2 mL | |
8 in. Color TFT LCD Touchscreen | |
Limited Warranty | |
USB, Wi-Fi, Ethernet | |
4°C/sec. (Block), 3°C/sec. (Sample) | |
18.3 lb. (8.5 kg) |