Learn More
Simport™ Scientific EasyDip™ Slide Staining Jars
User-friendly approach for staining microscope slides. These jars are a part of the EasyDip™ Slide Staining System. (See also the EasyDip Slide Staining Rack.)
Specifications
Material Acetal Plastic
For Use With (Application) Special stains, frozen sections and special processes
Height (English) 3.62 in.
Height (Metric) 92 mm
Slide Count 12 standard microscope slides
View More Specs
Description
Description
  • Vertical slide staining rack (sold separately) will hold up to 12 standard microscope slides
  • Slide writing area is kept out of staining fluid
  • Jars can be loosely joined to each other
  • Made of acetal plastic that resists attacks by most staining agents (except phenol, iodine or ferric chloride)
Specifications
Specifications
Acetal Plastic
3.62 in.
12 standard microscope slides
80 mL
Special stains, frozen sections and special processes
92 mm
Slide Staining Jar
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Simport™ Scientific EasyDip™ Slide Staining Jars