Learn More
Simport™ Scientific Stainless-Steel Base Molds
For applications in specimen embedding with Simport embedding rings and cassettes
Description
Description
  • Made of Stainless Steel
  • Highly polished surface
  • Superior thermal exchange
  • Well corners are rounded for optimal paraffin ribboning
For specimen embedding