missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIN-1 chloride, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
This compound (the active product of the prodrug SIN-10, molsidomine) acts as a vasodilator and inhibitor of platelet aggregation; longer acting than nitroprusside or nitroglycerin. Decreases myocardial necrosis and reperfusion-induced endothelial dysfunction in models of myocardial ischemia-reperfusion.
Specifications
Specifications
CAS | 16142-27-1 |
Molecular Formula | C6H11ClN4O2 |
Quantity | 50 mg |
Synonym | Amino-3-morpholinyl-1, 2, 3-oxadiazolium chloride |
Formula Weight | 206.63 |
Percent Purity | >98% |
Chemical Name or Material | SIN-1 chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'