Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
£66.95 - £560.00
Product Code | Brand | Description | For Use With (Equipment) | Holds | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Holds | Price | Quantity & Availability | |||
15886085
|
Thermo Scientific™
840-298900 |
Standard 10mm single-cell holder | GENESYS 140/150/180, BioMate 160 Spectrophotometers | 10 mm cuvette (1) |
£162.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15896085
|
Thermo Scientific™
840-299000 |
Beckman-style cell holder | GENESYS 140/150/180, BioMate 160 Spectrophotometers | Beckman-style short microcells |
£414.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15816095
|
Thermo Scientific™
840-303100 |
Long path cylindrical cell holder | GENESYS 140/150/180, BioMate 160 Spectrophotometers | 5 mm to 100 mm cylindrical cuvette (1) |
£186.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15826095
|
Thermo Scientific™
840-303200 |
Sample holder for film/filter accommodates filters/lenses up to 50 mm long, 80mm tall and 10mm thick. Comes assembled in a removable sample compartment liner. | GENESYS 140/150/180, BioMate 160 Spectrophotometers | Filter/lenses up to 50 mm long x 80 mm tall x 10 mm thick. Minimum filter size 18 mm. |
£278.00
Each |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15836095
|
Thermo Scientific™
840-303300 |
Disposable microcell holder | GENESYS 140/150/180, BioMate 160 Spectrophotometers | Microcells disposable 50 μL plastic Uvettes™ |
£423.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15866095
|
Thermo Scientific™
840-303800 |
Long path rectangular cell holder | GENESYS 140/150/180, BioMate 160 Spectrophotometers | 10 mm to 100 mm cuvette (1) |
£238.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11881762
|
Thermo Scientific™
335079-000 |
Thermostatted rectangular cell holder | SPECTRONIC 200 and GENESYS 30/40/50 Spectrophotometers | Square 10 mm Pathlength Cuvette |
£560.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15520592
|
Thermo Scientific™
840-277500 |
Test tube and vial holder accommodating 12 mm diameter up to 25 mm diameter test tubes and vials | GENESYS 30/40/50 Spectrophotometers | 12 mm to 25 mm diameter test tubes and vials |
£237.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15530592
|
Thermo Scientific™
840-277600 |
Test-tube cap covers test-tubes up to 15 cm/6 inches tall to allow measurements while the sample compartment lid is open. | GENESYS 30/40/50 Spectrophotometers | Tall tubes and vials (lid open) |
£157.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15540592
|
Thermo Scientific™
840-277700 |
Long path rectangular cell holder that accommodates 20-100 mm path length. Comes assembled in a removable compartment liner. | GENESYS 30/40/50 Spectrophotometers | 20 to 100 mm cuvette (1) |
£215.00
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15550592
|
Thermo Scientific™
840-277800 |
This sample accessory holds one cylindrical long path cell holder with 5-100 mm path length. Comes assembled in a removable sample compartment liner. | GENESYS 30/40/50 Spectrophotometers | 5 to 100 mm cylindrical cuvette (1) |
£177.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15570592
|
Thermo Scientific™
840-278100 |
Sample holder for film/filter accommodates filters/lenses up to 50 mm long, 80 mm tall and 6 mm thick. Comes assembled in a removable sample compartment liner. | GENESYS 30/40/50 Spectrophotometers | Filter/lenses up to 50 mm long x 80 mm tall x 6 mm thick |
£313.00
Each |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15580592
|
Thermo Scientific™
840-278200 |
Accessory Kit for Hose Entry/Exit | GENESYS 30/40/50 Spectrophotometers | Entry and exit hoses (included) |
£237.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11751151
|
Thermo Scientific™
335911-000 |
Cylindrical cell holder | 840-303600 four-position carousel base | 5 - 100 mm cylindrical cells |
£178.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11771151
|
Thermo Scientific™
336028-000 |
10 mm Square Cell Holder | GENESYS 30/40/50 Spectrophotometers | Single-cell cuvette or 10 mm diameter test tubes |
£66.95
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15520602
|
Thermo Scientific™
840-278600 |
Standard 10 mm single-cell holder | GENESYS 30/40/50 Spectrophotometers | 10 mm cuvette (1) |
£153.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11893744
|
Thermo Scientific™
335917000 |
Adjustable filter holder | GENESYS 30/40/50 Spectrophotometers | Filters up to 10 mm thick |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Beschreibung
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.