Learn More
Socorex™ Acura™ Single-channel Macropipette Volumetric Module
Increase the versatility of your Acura™ electronic macropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ Acura™ Single-channel Macropipette Volumetric Module is an excellent and easily attachable/detachable choice for your single-channel macropipetting needs.
Brand: Socorex™ 800.5000
This item is not returnable.
View return policy
Description
- One of 27 available volumetric assemblies.
- Increase versatility and save costs.
- Disassembles and reassembles without tools.
- Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
Acura™ Electro™ 926 MicroPipette | |
Acura™ | |
0.25 to 5 mL | |
1 Each |
1 | |
Electronic Pipette Volumetric Module | |
Acura™ Electro 926 Micropipette |