Learn More
Socorex™ Acura™ Single-channel Micropipette Volumetric Module
Increase the versatility of your Acura™ electronic micropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ Acura™ Single-channel Micropipette Volumetric Module is an excellent and easily attachable/detachable choice for your single-channel micropipetting needs.
Brand:  Socorex™ 800.0010
 View more versions of this product
Product Code. 11743276
  • £76.60 / Each
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One of 27 available volumetric assemblies.
    • Increase versatility and save costs.
  • Disassembles and reassembles without tools.
  • Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
Specifications
Acura™ Electro™ 926 MicroPipette
Acura™
0.5 to 10 μL
1 Each
1
Electronic Pipette Volumetric Module
Acura™ Electro 926 Micropipette