missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Single drawer unit, blue, 140 x 410 x 430mm
Brand: Manutan A349065
This item is not returnable.
View return policy
Description
All steel construction drawers for below workbench. Lockable cupboard and drawers, giving secured storage.- Blue
- Maximum load per drawer: 25kg
- Dimensions (H x W x D): 140mm x 410mm x 430mm