Learn More
Hughes Single Nozzle Handheld Eye/Face/Body Wash
A wall mounted, handheld eye, face and body wash with one nozzle. Developed to suit hazardous environments where there is a risk of small spillages of chemicals or dangerous substances.
Brand:  Hughes HEWS5021GB
Product Code. 17772338
  • £374.67 / Each
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A wall mounted, handheld eye, face and body wash with one nozzle
  • Developed to suit hazardous environments where there is a risk of small spillages of chemicals or dangerous substances
  • 5 m of reinforced hose makes these handheld units particularly useful for treating disabled personnel
  • The lock-on actuating button for ease of use meets European and international standards
  • Fitted with dust covers, a non-return valve and a flow regulator
Specifications
Specifications
Red
10.5 cm
Eye, Face and Body Combination Wash Stations
7 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hughes Single Nozzle Handheld Eye/Face/Body Wash > Eye/Face/Body Wash; Color: Red