missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ SNP Genotyping Assay, human

Product Code. 10658265 Shop All Applied Biosystems Products
Click to view available options
Quantity:
L (2400 reactions), made to order
M (1000 reactions), made to order
S (300 reactions), made to order
Unit Size:
300 reactions
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10658265

Brand: Applied Biosystems™ 4351384

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Two unlabeled PCR primers: (forward and reverse primers at 900nM final concentration); One Applied Biosystems VIC™ dye: MGB labeled probe detects the Allele 1 sequence (probes at 200nM final concentration); One Applied Biosystems 6FAM™ dye: MGB labeled probe detects the Allele 2 sequence (probes at 200nM final concentration)

5' nuclease assay to discriminate between two alleles of a specific SNP for use in genotyping studies

  • Large selection of SNP assays advances validation and screening studies for disease association and population stratification applications
  • Easy-to-use, single-tube format
  • Robust assay design provides highly accurate, reproducible and reliable results
  • Simple workflow enables quick results

  • Large selection of SNP assays advances validation and screening studies for disease association and population stratification applications
  • Easy-to-use, single-tube format
  • Robust assay design provides highly accurate, reproducible and reliable results
  • Simple workflow enables quick results

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Two unlabeled PCR primers: (forward and reverse primers at 900nM final concentration); One Applied Biosystems VIC™ dye: MGB labeled probe detects the Allele 1 sequence (probes at 200nM final concentration); One Applied Biosystems 6FAM™ dye: MGB labeled probe detects the Allele 2 sequence (probes at 200nM final concentration). Store at -15° to -25°C
Description Single-Tube TaqMan SNP Genotyping Assays
Quantity S (300 reactions), made to order
For Use With (Application) Genotyping
No. of Reactions 300 reactions
Sample Type Mouse

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.