missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ SNP Genotyping Assay, human
GREENER_CHOICE

Product Code. p-8015468 Shop All Applied Biosystems Products
Click to view available options
Quantity:
L (2400 reactions), made to order
M (1000 reactions), made to order
S (300 reactions), made to order
Unit Size:
300 reactions
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10167144

Brand: Applied Biosystems™ 4351374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Two unlabeled PCR primers: (forward and reverse primers at 900nM final concentration); One Applied Biosystems VIC™ dye: MGB labeled probe detects the Allele 1 sequence (probes at 200nM final concentration); One Applied Biosystems 6FAM™ dye: MGB labeled probe detects the Allele 2 sequence (probes at 200nM final concentration)

5' nuclease assay to discriminate between two alleles of a specific SNP for use in genotyping studies

  • Large selection of SNP assays advances validation and screening studies for disease association and population stratification applications
  • Easy-to-use, single-tube format
  • Robust assay design provides highly accurate, reproducible and reliable results
  • Simple workflow enables quick results

Applied Biosystems TaqMan SNP Genotyping Assays use TaqMan 5'‐nuclease chemistry to amplify and detect specific polymorphisms in purified genomic DNA samples. Each assay enables genotyping of individuals for a single nucleotide polymorphism (SNP) and consists of two sequence-specific primers and two TaqMan minor groove binder (MGB) probes with non-fluorescent quenchers (NFQ). One probe is labeled with VIC dye to detect the Allele 1 sequence; the second probe is labeled with FAM dye to detect the Allele 2 sequence.

Our predesigned TaqMan SNP Genotyping Human Assays are a genome-wide collection of millions of human assays, including common 1,000 Genome SNPs, HapMap SNPs, and coding SNPs.

Benefits:

  • Proven—gold-standard TaqMan chemistry and robust assay designs deliver accurate, reproducible, and reliable results
  • Easy—convenient single-tube format and simple workflow provide an easy path to trusted results; no optimization required
  • Relevant—extensive collection of predesigned human assays offers direct access to content that is relevant to your research
  • Tested—all human SNP genotyping assays are functionally tested to ensure allelic discrimination

Approximate ship time

4–6 days in North America and 6–10 days in Europe

TaqMan SNP Genotyping Assays require only three reaction components for PCR: purified genomic DNA (1–20 ng), the assay solution, and TaqMan Genotyping Master Mix (or another compatible master mix) (sold separately).

All assay designs are the product of our bioinformatics pipeline, optimized over the course of more than a decade by leveraging manufacturing and assay performance data. TaqMan Assays have been cited in over 40,000 publications and are backed by more than 350 patents.

All of our predesigned TaqMan Assays are covered by the TaqMan Assays qPCR Guarantee.*

Recommended master mix (sold separately): TaqMan Genotyping Master Mix (Cat. No. 4371355)

*Terms and conditions apply. For complete details, go to www.thermofisher.com/taqmanguarantee.

TRUSTED_SUSTAINABILITY

Specifications

3'Primer Modification None
5'Primer Modification None
Concentration 80X
Content And Storage 1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers).

Store at -15 to -25°C.

Description VIC™ - MGB probe detects Allele 1, FAM - MGB probe detects Allele 2
Form Frozen
For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ Absolute Q Digital PCR System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex ProFlex PCR System*, VeritiPro*, SimpliAmp*, MiniAmp*, Automated Thermal Cycler** If a thermal cycler is used for PCR amplification, the optional pre-read and the post-read must be performed separately on a real-time PCR system in order to detect and record fluorescent signals.
Green Features Sustainable packaging
Internal Probe Modification VIC™ (5'), FAM (5'), MGB (Minor Groove Binder) (3')
Label or Dye FAM, VIC
Product Line TaqMan
Purification Method Solid Phase Extraction (SPE)
Purity or Quality Grade RNase-Free, DNase-Free
Quantity L (2400 reactions), made to order
Shipping Condition Room Temperature
Species Human
Target Over 4.5 million genome-wide human assays are available
For Use With (Application) Genotyping
No. of Reactions 2400 reactions
Sample Type Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.