Learn More
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly safely transfers, samples, and stores biopharmaceutical products and reagents
Brand: Masterflex™ 95644-28
This item is not returnable.
View return policy
Description
- Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
- Assemblies are gamma irradiated - eliminates labor and validation costs associated with assembly and cleaning
Specifications
Gamma Irradiated | |
2 L |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly > 2000 mL
Spot an opportunity for improvement?Share a Content Correction