Learn More
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly safely transfers, samples, and stores biopharmaceutical products and reagents
Brand:  Masterflex™ 95644-24
 View more versions of this product
Product Code. 16699561
  • £154.00 / Each
Explore more special offers
Description
Description
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
  • Assemblies are gamma irradiated - eliminates labor and validation costs associated with assembly and cleaning
Specifications
Specifications
Gamma Irradiated
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly > 500 mL