Learn More
Poulten Graf™ Single Use Luer-Tip Two-Part Syringe
Use these syringes for veterinary medicine applications. Poulten Graf™ Single Use Luer-Tip Two-Part Syringe are PVC-free and non-toxic, sterile packed and depatched in blister packs. The single-use syringes are extremely tight, and have a transparent, smooth operating cylinder.
Specifications
Autoclavable Autoclavable
Certifications/Compliance CE
Material Polypropylene
Description Screen printed graduation
Product Type Luer-tip 2-part Syringe
View More Specs
Description
Description
  • Produced in accordance with DIN 13098
  • Available in a variety of sizes, with and without needle
Specifications
Specifications
Autoclavable
Polypropylene
Luer-tip 2-part Syringe
Luer Tip
Sterile
CE
Screen printed graduation
Single-use
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Poulten Graf™ Single Use Luer-Tip Two-Part Syringe