missing translation for 'onlineSavingsMsg'
Learn More

Manutan Single wall box, 305mm x 229mm x 305mm

Brand:  Manutan A329263

Encompass_Preferred

Product Code. 12858358

  • £24.81 / Pack of 25
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The cost-effective solution for packing less fragile items, economical and ideal for every day use. Strong, single wall boxes with brown kraft finish. Supplied flat for ease of storage.
  • Internal dimensions (L x H): 305mm x 305mm
  • Load capacity: 10kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers