missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Single Well High Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
Brand: Axygen™ RES-SW96-HP-SI
193.51 GBP valid until 2024-11-15
Use promo code "21643" to get your promotional price.
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
240 mL | |
Sterile | |
Individually Wrapped; 25/Cs. | |
Single-use |
Clear | |
Polypropylene | |
Reagent Reservoir | |
96 |