missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Single Well Low Profile Reagent Reservoirs
GREENER_CHOICE

Manufactured with virgin polypropylene for excellent chemical resistance

Brand:  Axygen™ RES-SW384-LP

289.75 GBP valid until 2024-12-31
Use promo code "22009" to get your promotional price.


 View more versions of this product

Product Code. 11330275

  • £322.00 / Pack of 25
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANSI/SLAS, RNase/DNase-free, non-pyrogenic
92 mL
Non-sterile
25/Cs.
Single-use
Clear
Polypropylene
Reagent Reservoir
384
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.