missing translation for 'onlineSavingsMsg'
Learn More

IL6R Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 365) of human IL6R (NP_000556.1) fused to a polyhistidine tag at the C-terminus

£68.50 - £1105.00

Specifications

For Use With (Application) Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
Gene ID (Entrez) 3570
Molecular Weight (g/mol) 40kDa
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13485866
View Documents Promotion Details
Invitrogen
10398H08H5
5μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13495866
View Documents Promotion Details
Invitrogen
10398H08H50
50μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13475866
View Documents Promotion Details
Invitrogen
10398H08H250
5 x 50μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Interleukin 6 receptor (IL6R), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human IL6 at 8μg/mL (100μL/well) can bind recombinant human IL6R with a linear range of 1.25-20.0ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
40kDa
Truncated/Recombinant
IL6R, CD126, IL-6R-1, IL-6R-alpha, IL6RA, mgC104991
IL6R
Mammalian
Lyophilized
≥90% by SDS-PAGE
3570
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Interleukin 6 receptor
His Tag
Cytokines & Receptors/CD Proteins
Interleukins, Cytokine Receptors
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.