missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus™ Six Block Analog Dry Block Heater HB6AL
Click to view available options
Description:
Dry Block Heater, 6 Block, HB6AL, EU
Description
- These high wattage, constant-temperature dry block heaters are economical, versatile and compact
- Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures
- Each of the five models accept separate interchangeable modular blocks, accommodating various sample containers such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes
- Each block has a thermometer well for measuring block temperature
- Anodised aluminum modular blocks provide superior temperature stability and heat transfer
- Heaters require OHAUS modular heating blocks for operation
- Analogue controls
Specifications
Specifications
Certifications/Compliance | TUV |
Controller Type | Analog |
Description | Dry Block Heater, 6 Block, HB6AL, EU |
Temperature Range (Metric) | 5°C to 100°C (Low), 75°C to 150°C (High) |
Type | Dry Bath Block |
Heat Up Time | 75 min |
Model | HB6AL |
No. of Blocks | 6 |
Temperature Stability | ±2°C |
Temperature Uniformity | ±0.1°C at 37°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'