Learn More
Masterflex™ Six Channel Syringe Pump
Six-channel pump can both withdraw and infuse
Brand:  Masterflex™ 75900-55
3841.64 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 11759889
  • £5160.00 / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Programming features:
    • Programming commands allow dispensing of a specified amount, automatic rates changes, direction changes, programmed pauses, external or user input, and sending or receive signals from other equipment.
    • Non-volatile memory restores all setup parameters and the pumping program upon turning the pump on.
    • An audible buzzer can be programmed to alert when an alarm condition occurs or the pumping program completes.
    • Syringe purge mode will infuse or refill syringe at top speed to purge air from the line or to fill the syringe.
    • Versatile pumping features allow infusion or withdrawal in μl/hr, μl/min, ml/hr, and mL/min.
  • Communication features:
    • Network up to 100 pumps from a single communications port on a computer.
    • Network communication protocols allow the ability to send a network burst transmission simultaneously to 10 pumps in the network with no lag, resulting in consistent laboratory setups.
    • Optional communications protocol utilizes error checking to ensure completely reliable signal transmissions by eliminating errors caused by disconnections or electromagnetic interference.
Specifications
Specifications
230 V
22.3 mL/min.
15.25 in. X 10.5 in. X 4.75 in.
6 x 60 mL