missing translation for 'onlineSavingsMsg'
Learn More

Manutan Six tier commercial locker, grey/grey, 1800mm x 300mm x 500mm

Brand:  Manutan A330052

Encompass_Preferred

Product Code. 12894869

  • £229.90 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ideal for commercial, industrial, educational and leisure environments. 300mm deep lockers, fitted with hat shelf and two coat hooks. 500mm deep lockers, fitted with hat shelf, coat rail and single coat hook. Welded steel construction for added strength and rigidity. Supplied with cylinder lock and two keys per compartment. Pre-drilled for nesting side by side and/or back to back. Powder coated finish.
  • Grey door
  • Dimensions (H x W x D): 1800mm x 300mm x 500mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.