Learn More
Tocris Bioscience™ SJ 1008066
MAGE-A11 inhibitor disrupts MAGE-A11:PCF11 interaction
Brand:  Tocris Bioscience™ 7542/5
Additional Details : Weight : 0.00970kg
Product Code. 18075252
  • £253.43 / 5mg
Estimated Shipment: 12-08-2024
to see stock.
Description
Description
SJ 1008066 is a cell permeable inhibitor of testis-restricted melanoma antigen protein MAGE-A11 (IC50 = 0.13 μM). SJ 1008066 binds to the MAGE homology domain (MHD) and disrupts MAGE-A11:PCF11 interaction. SJ1008066 exhibits no effect on MAGE-A3:AMPKα1 or MAGE-F1:MMS19 interactions in TR-FRET and cell lysate pulldown assays.
Specifications
Specifications
N1-((1 H-Indol-5-yl)methyl)-N3-(quinolin-4-yl)propane-1,3-diamine
Ubiquitin Ligase (E3) Inhibitors
98%
5 mg
C21H22N4