missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKF 38393 hydrobromide, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Prototypical D1-like dopamine receptor selective partial agonist (Ki values are 1, ∽ 0.5, ∽ 150, ∽ 5000 and ∽ 1000 nM for D1, D5, D2, D3 and D4 receptors respectively).
Specifications
CAS | 20012-10-6 |
Molecular Formula | C16H18BrNO2 |
Quantity | 100 mg |
Synonym | skf 38393 hydrobromide, 1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, 1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diolhydrobromide, +/--1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, 1h-3-benzazepine-7,8-diol,2,3,4,5-tetrahydro-1-phenyl, 1-phenyl-2,3,4,5-tetrahydro-1h-benzo d azepine-7,8-diol hydrobromide, 7,8-dihydroxy-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrobromide |
InChI Key | INNWVRBZMBCEJI-UHFFFAOYSA-N |
SMILES | C1CNCC(C2=CC(=C(C=C21)O)O)C3=CC=CC=C3.Br |
IUPAC Name | 5-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol;hydrobromide |
Molecular Weight (g/mol) | 336.229 |
PubChem CID | 12928470 |
Percent Purity | >99% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'