missing translation for 'onlineSavingsMsg'
Learn More

SKOR2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA561186

Product Code. 15779001

  • £427.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: MASSPLPGPN DILLASPSSA FQPDTLSQPR PGHANLKPNQ VGQVILYGIP IVS Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Exhibits transcriptional repressor activity. Acts as a TGF-beta antagonist in the nervous system.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SKOR2
Polyclonal
Unconjugated
SKOR2
CH18515; Corl2; EG664805; functional smad suppressing element 18; functional Smad suppressor element on chromosome 18; functional Smad-suppressing element on chromosome 18; FUSSEL18; Fussel-18; fussel-18 homolog; Gm7348; ladybird homeobox corepressor 1-like protein; LBX1 corepressor 1-like protein; SKI family transcriptional corepressor 2; Skor2; Transcriptional corepressor Corl2
Rabbit
Antigen affinity chromatography
RUO
664805
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
A7M7C7
SKOR2
Recombinant protein corresponding to Human SKOR2
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only