missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB

Safe and approved storage of aggressive, non-flammable hazardous materials in working areas.

Brand:  asecos™ 3062289230624

Product Code. 13479716

  • POA / Each

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers

Alert:

For indoor use only

This item is not returnable. View return policy

Description

Description

Safe and approved storage of aggressive, non flammable hazardous materials in working areas

Function / construction

  • Robust construction and longevity: made out of highly resistant melamin resin special plates
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • No unauthorised use: door lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Certificates

  • type tested in accordance with EN 14727
  • EN 16121/16122 conformity
  • GS certified
Specifications
Show More
Product Suggestions

Product Suggestions

Videos

Videos

  • Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      asecos SL-LINE SL-CLASSIC safety storage cabinets

    SDS
    Documents Certificates
    View accessory products
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    asecos™ Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB > 570 x 590 x 600mm, 1 x pull-out shelf (panel melamine resin-coated/PP)

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.