missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB
Safe and approved storage of aggressive, non-flammable hazardous materials in working areas.
Brand: asecos™ 3062289230624
Alert:
For indoor use only
Description
Safe and approved storage of aggressive, non flammable hazardous materials in working areas
Function / construction
- Robust construction and longevity: made out of highly resistant melamin resin special plates
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- No unauthorised use: door lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Certificates
- type tested in accordance with EN 14727
- EN 16121/16122 conformity
- GS certified
Specifications
DIN EN 16121/16122 conformity, GS certified | |
570 mm | |
Under bench cabinet for acids and alkalis SL-CLASSIC-UB model SL.060.059.UB.T light grey, interior equipment with 1 x pull-out shelf (panel/PP) | |
570 x 590 x 600 mm | |
Safe and approved storage of aggressive, non fla mmable hazardous materials in working areas | |
562 mm | |
1 | |
Pull-out | |
590 mm | |
Light Gray | |
Under bench cabinet for acids and alkalis |
1 x pull-out shelf (panel melamine resin-coated/PP) | |
531 mm | |
531 x 489 x 562 mm | |
Wing, Left-hinged | |
600 mm | |
SL.060.059.UB.T | |
1 | |
Melamine resin laminated | |
485 mm | |
Chipboard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
asecos™ Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB > 570 x 590 x 600mm, 1 x pull-out shelf (panel melamine resin-coated/PP)
Spot an opportunity for improvement?Share a Content Correction