missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ SL-CLASSIC-UB Underbench Safety Storage Cabinets with 2 Doors

For the safe and proper storage of aggressive, non-flammable hazardous materials in working areas underneath work benches and fume cupboards.

Brand:  Asecos™ 3320089230620

887.12 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : NEW

Additional Details : Weight : 60.00000kg

 View more versions of this product

Product Code. 15648845

  • £988.98 / Each
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

  • Robust construction and longevity – underbench cabinets completely made of melamine resin and flame retardant special plates
  • Safe and separate storage – acids and alkalis can be stored separately
  • Two hermetically divided storage compartments, integrated aeration and venting system, corrosion resistance of the metal-free air ducts
  • In accordance with TRGS 510
  • Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
  • Interior equipment available includes pull-out shelves and bottom collecting sumps
  • No unauthorized use – doors lockable with profile cylinder (integration into an existing locking system possible)
  • Ventilation – integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Specifications

Specifications

570 x 1100 x 800 mm
530 x 489 x 760 mm
2 x 25 kg/11 L PP pull-out shelves with PP sump
Storage of aggressive and non-flammable hazardous substances
Wing
2
94 kg
Light Gray
Safety storage cabinet
2 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg
Melamine Resin
TRGS 510
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers