missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ SL-CLASSIC-UB Underbench Safety Storage Cabinets with 2 Doors
For the safe and proper storage of aggressive, non-flammable hazardous materials in working areas underneath work benches and fume cupboards.
£988.98 - £1215.82
Specifica
Certifications/Compliance | TRGS 510 |
---|---|
Color | Light Gray |
Material | Melamine Resin |
Product Type | Safety storage cabinet |
Door Style | Wing |
Codice del prodotto | Marca | Load Bearing Capacity | Dimensions (D x W x H) | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Load Bearing Capacity | Dimensions (D x W x H) | Prezzo | Quantità e disponibilità | ||||
15411583
|
Asecos™
3320089230621 |
4 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg | 570 x 1100 x 800 mm |
£1215.82
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15648845
|
Asecos™
3320089230620 |
2 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg | 570 x 1100 x 800 mm |
£988.98
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15984340
|
Asecos™
3061989230621 |
4 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg | 570 x 1100 x 600 mm |
£1179.78
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Descrizione
- Robust construction and longevity – underbench cabinets completely made of melamine resin and flame retardant special plates
- Safe and separate storage – acids and alkalis can be stored separately
- Two hermetically divided storage compartments, integrated aeration and venting system, corrosion resistance of the metal-free air ducts
- In accordance with TRGS 510
- Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
- Interior equipment available includes pull-out shelves and bottom collecting sumps
- No unauthorized use – doors lockable with profile cylinder (integration into an existing locking system possible)
- Ventilation – integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Specifica
TRGS 510 | |
Melamine Resin | |
Wing | |
2 |
Light Gray | |
Safety storage cabinet | |
Storage of aggressive and non-flammable hazardous substances |