Learn More
Asecos™ SL-CLASSIC-UB Underbench Safety Storage Cabinets with 3 Doors
For the safe and proper storage of aggressive, non-flammable hazardous materials in working areas underneath work benches and fume cupboards.
Brand: Asecos™ 3061689230617
1112.19 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 100.00000kg
Description
- Robust construction and longevity – underbench cabinets completely made of melamine resin and flame retardant special plates
- Safe and separate storage – acids and alkalis can be stored separately
- Two hermetically divided storage compartments, integrated aeration and venting system, corrosion resistance of the metal-free air ducts
- In accordance with TRGS 510
- Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
- Interior equipment available includes pull-out shelves and bottom collecting sumps
- No unauthorized use – doors lockable with profile cylinder (integration into an existing locking system possible)
- Ventilation – integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Specifications
570 x 1400 x 600 mm | |
531 x 489 x 562 mm | |
2 x 25 kg/11 L PP pull-out shelves with PP sump | |
Storage of aggressive and non-flammable hazardous substances | |
Wing | |
3 |
93.5 kg | |
Light Gray | |
Safety storage cabinet | |
2 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg | |
Melamine Resin | |
TRGS 510 |