missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ SL-CLASSIC-UB Underbench Safety Storage Cabinets with 3 Doors
For the safe and proper storage of aggressive, non-flammable hazardous materials in working areas underneath work benches and fume cupboards.
£1206.28 - £1433.12
Specifications
Certifications/Compliance | TRGS 510 |
---|---|
Color | Light Gray |
Dimensions (D x W x H) | 570 x 1400 x 600 mm |
Dimensions (D x W x H) Interior | 531 x 489 x 562 mm |
Material | Melamine Resin |
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
15955770
|
Asecos™
3061689230618 |
4 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg |
£1433.12
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15874178
|
Asecos™
3061689230617 |
2 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg |
£1206.28
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Robust construction and longevity – underbench cabinets completely made of melamine resin and flame retardant special plates
- Safe and separate storage – acids and alkalis can be stored separately
- Two hermetically divided storage compartments, integrated aeration and venting system, corrosion resistance of the metal-free air ducts
- In accordance with TRGS 510
- Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
- Interior equipment available includes pull-out shelves and bottom collecting sumps
- No unauthorized use – doors lockable with profile cylinder (integration into an existing locking system possible)
- Ventilation – integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Specifications
TRGS 510 | |
570 x 1400 x 600 mm | |
Melamine Resin | |
Wing | |
3 |
Light Gray | |
531 x 489 x 562 mm | |
Safety storage cabinet | |
Storage of aggressive and non-flammable hazardous substances |