missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB
Under bench cabinet for acids and alkalis with pull out shelf.
Brand: asecos™ 30619-001-30621
Alert:
For indoor use only
Description
- Robust construction and longevity: made out of highly resistant melamin resin special plates
- User-friendly: access to all containers, 135° opening of the wing door, interior of the cabinet completely visible
- No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Specifications
GS approval, CE Conformity, EN 14727 Conformity | |
531 mm | |
Wing | |
600 mm | |
SL.060.110.UB.2T | |
60 kg | |
489 mm | |
Steel |
4 x pull-out shelf (panel/PP) | |
531 x 489 x 562 mm | |
Safe and approved storage of aggressive, non flammable hazardous materials in working areas | |
531 mm | |
Powder-coated | |
1100 mm | |
Light Gray | |
Under bench cabinet for acids and alkalis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction