missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB
Under bench cabinet for acids and alkalis with pull out shelf.
Brand: asecos™ 3062389230625
Alert:
For indoor use only
Description
Safe and approved storage of aggressive, non flammable hazardous materials in working areas
Function / construction
- Robust construction and longevity: made out of highly resistant melamin resin special plates
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- No unauthorised use: door lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Certificates
- type tested in accordance with
- EN 16121/16122 conformity
- GS certified
Specifications
DIN EN 16121/16122 conformity, GS certified | |
570 mm | |
Pull-out shelf is panel melamine resin-coated/PP | |
570 x 590 x 600 mm | |
Safe and approved storage of aggressive, non flammable hazardous materials in working areas | |
562 mm | |
SL.060.059.UB.TR | |
2 | |
Melamine resin laminated | |
590 mm | |
Light Gray | |
Acids and Alkalis Cabinet (Underbench) |
2 Pull-out Shelves, Sump | |
531 mm | |
531 x 488 x 562 mm | |
Wing, Right-hinged | |
600 mm | |
25 kg/11 L | |
1 | |
Pull-out | |
53.50 kg | |
488 mm | |
Chipboard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
asecos™ Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB > Includes: 2 x pull-out shelf with PP-sump, load capacity: 25 kg, capacity: 11 litres (panel melamine resin-coated/PP)
Spot an opportunity for improvement?Share a Content Correction