Learn More
Thermo Scientific™ SL4FR Plus-MD, 208-240V TX-1000 Blood Tube Package
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol.
Brand: Thermo Scientific™ 75016064
Description
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol. Exceptional capacity to process 196x 5 mL blood tubes in one run, under biocontainmenyt certified lids. Contains centrifuge, TX-1000 rotor, buckets, biocontainment lids, 5/7 mL blood tube adapter (maximum capacity 196 x 5/7 mL), 10 mL blood tube adapter (maximum capacity 148 x 10 mL).
-
Easy to Use:
- Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
- Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
- Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
Thermo Scientific™ Auto-Lock™ III Rotor System:
- Secure locking system allows easy push-button installation and exchange of rotors.
- System saves time when switching between applications.
- Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
Thermo Scientific™ ClickSeal™ Bucket Sealing System:
- Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
- System has glove-friendly, one-handed open/close capability.
- System is certified by CAMR™ in Porton Down, UK.
Thermo Scientific™ SMARTSpin™ Technology:
- Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
- Technology optimizes safety and improves separations.
Space-Saving Design:
- Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
Energy Efficient:
- System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Specifications
Direct, Brushless induction low profile motor with choices of 9 acceleration, 10 deceleration rates | |
Part Numbers In Pk. 75009973 (centrifuge),75003017(rotor),75003001(set of buckets),75007309(lids),75003671(set of adapters),75003697 75003672(set of adapters) | |
General Purpose Centrifuge Series |