Learn More
Bollé Safety Slam+ Platinum™ CSP Safety Glasses
Description
- Recommended for extremely hot or extremely cold environments
- Comfort, twin material black and orange temples
- All-round vision
- PLATINUM coating
Specifications
Specifications
Certifications/Compliance | Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures - T |
Lens Coating | Anti-fog, Scratch-resistant |
Frame Color | Black, Orange |
Temple Style | Twin-material |
Temple Color | Black |