missing translation for 'onlineSavingsMsg'
Learn More

SLC10A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Bio-Techne NBP1-88813

 View more versions of this product

Product Code. 18254476

  • £392.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SLC10A2 Polyclonal specifically detects SLC10A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SLC10A2
Polyclonal
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Apical sodium-dependent bile acid transporter, ASBT, IBAT, ileal apical sodium-dependent bile acid transporter, Ileal Na(+)/bile acid cotransporter, ileal sodium/bile acid cotransporter, Ileal sodium-dependent bile acid transporter, ISBT, Na(+)-dependent ileal bile acid transporter, NTCP2PBAM, Sodium/taurocholate cotransporting polypeptide, ileal, solute carrier family 10 (sodium/bile acid cotransporter family), member 2, Solute carrier family 10 member 2
Rabbit
Affinity Purified
RUO
6555
Human
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
SLC10A2
This antibody was developed against Recombinant Protein corresponding to amino acids:NKAEIPESKENGTEPESSFYKANGGFQPDE
0.1 mL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only