missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A6 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111912
Additional Details : Weight : 0.01000kg
Description
SLC39A6 (extracellular) Polyclonal antibody specifically detects SLC39A6 (extracellular) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
SLC39A6 (extracellular) | |
Polyclonal | |
Unconjugated | |
SLC39A6 | |
EMBL:AAH97493.1}; endoplasmic reticulum membrane protein; Endoplasmic reticulum membrane-linked protein; Ermelin; Estrogen-regulated protein LIV-1; LIV1; LIV-1; LIV-1 protein, estrogen regulated; Slc39a6; slc39a6 {ECO:0000312; solute carrier family 39 (metal ion transporter), member 6; solute carrier family 39 (zinc transporter), member 6; Solute carrier family 39 member 6; Zinc transporter ZIP6; ZIP6; ZIP-6; Zrt- and Irt-like protein 6 | |
Peptide corresponding to amino acid residues 251-264 of mouse ZIP6 (Extracellular, N-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q4V887, Q8C145, Q13433 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25800, 106957, 291733 | |
-20°C | |
Lyophilized |