missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC4A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534207
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey (100%); Marmoset, Elephant, Horse (88%); Panda, Bat (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Plasma membrane anion exchange protein of wide distribution.Specifications
SLC4A2 | |
Polyclonal | |
Unconjugated | |
SLC4A2 | |
AE 2; AE2; Aep2; anion exchange protein 2; anion exchanger 2; anion exchanger 2 type a; anion exchanger 2 type b1; anion exchanger 2 type b2; B3RP; B3rp2; B3RP-2; Band 3-related protein; band 3-related protein 2; BND3L; EPB3L1; Erythrocyte band 3; erythrocyte membrane protein band 3-like 1; HKB3; MPB3L; NBND3; non-erythroid band 3-like protein; Slc4a2; solute carrier family 4 (anion exchanger), member 2; solute carrier family 4 member 2; Solute carrier family 4, member 2, anion exchange protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6522 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.11 mg/mL | |
PBS with 0.1% sodium azide | |
P04920 | |
SLC4A2 | |
Synthetic 16 amino acid peptide from internal region of human SLC4A2. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction