missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC9A9 Mouse anti-Human, Mouse, Rat, Clone: 1B8D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66577-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.Specifications
SLC9A9 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BZ00, Q8IVB4 | |
Slc9a9 | |
SLC9A9 Fusion Protein Ag4660 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1B8D9 | |
Unconjugated | |
SLC9A9 | |
Na(+)/H(+) exchanger 9, Nbla00118, NHE 9, NHE9, SLC9A9, Sodium/hydrogen exchanger 9 | |
Mouse | |
Protein A | |
RUO | |
285195, 331004, 363115 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction