missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Sleeve Groove

Product Code. 10598461
Click to view available options
Quantity:
1 Each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10598461

Brand: Bohlender™ H93014

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these reusable PTFE sleeve grooves with rotary evaporators. Bohlender™ BOLA™ Sleeve Grooves feature solid construction for continued use and easily accessible, easy to clean sealing rings.

  • Made of PTFE, with sealing rings (“ribs”) on the outside
  • Without gripping ring and therefore usable with joint clamps
  • For vacuum up to 0.1 mbar
  • Temperature range: -200 to +250°C
  • Universal chemical resistance
  • Vacuum suitable

Specifications

For Use With (Equipment) Rotary evaporator
Material PTFE
Product Type Sleeve Grooves
Quantity 1 Each
Temperature Range (Metric) -200°C to +250°C
Standard Taper Joint No. NS 71/51
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.