Learn More
Bohlender™ Sleeve Grooves With Ribs
Product Code. 10629031
Click to view available options
Quantity:
1/Pack
Unit Size:
Each
Product Code. 10629031
Brand: Bohlender™ H93210
Only null left
Add to basket
Add to basket
Specifications
For Use With (Equipment) Rotary evaporator
Material PTFE
Product Type Sleeve Grooves With Ribs
Quantity 1/Pack
Temperature Range (Metric) -200°C to +250°C
Standard Taper Joint No. NS 45/50