missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Devices, 3.5K MWCO

Perform sample buffer exchange with these 3500-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.

Brand:  Thermo Scientific™ 69558

75.26 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11899410

  • £106.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Slide-A-Lyzer MINI Dialysis Devices, 3.5K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules >3.5kDa in volumes from 10 μL to 2 mL. They were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices.

Slide-A-Lyzer MINI Dialysis Devices are manufactured in a clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use. These 3.5K MWCO dialysis devices are available in three sizes (0.1, 0.5, and 2 mL capacities), and are designed to allow easy sample addition and removal using a standard laboratory pipette. The MINI devices are effective and convenient for accomplishing typical sample desalting or buffer exchange in 4–8 hours. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters, and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with common chemicals and buffers.

Features of Slide-A-Lyzer MINI Dialysis Devices, 3.5K MWCO:
Three convenient sizes—dialysis devices for 0.1, 0.5, and 2 mL samples fit standard 1.5-mL micro centrifuge, 15- and 50-mL conical tubes, respectively
Excellent sample recovery—low-binding plastic and small membrane surface area minimize sample loss
One-step protocol—pipette sample into the MINI device and place in tube containing the dialysis buffer; no laborious assembly, device preparation, or expensive equipment are required
100% leak-tested—innovative design does not permit ‘wicking’ that can occur in home-made devices
Time saving—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes, which improves sample resolution, quantification, and conjugation

The 0.5- and 2-mL MINI dialysis devices fit in and are capped by inserting them into the included 15- and 50-mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI dialysis devices, low molecular weight contaminant removal and buffer exchange and desalting can be accomplished within 4–8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.

The 0.1 mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL micro-centrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker.

Using the 0.1 mL MINI Device, 100 μL of pH 2.8 glycine buffer can be equilibrated to 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes. As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cup, Microcentrifuge Kit
3.5 kDa
MINI Dialysis Device
Buffer Exchange, Protein
10 Samples
Store at room temperature
Slide-A-Lyzer™
Cellulose Membrane, Dialysis
MINI Dialysis Device Kit
0.1 mL
10 Devices
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.