Learn More
Thermo Scientific™ Slide-A-Lyzer™ Accessory Float Buoys and Syringes
Slide-A-Lyzer Float Buoys and Syringes are available to support a variety of dialysis cassettes for volumes of 0.1 to 20 mL.
£41.73 - £53.00
Specifications
Content And Storage | Store at room temperature. |
---|---|
Membrane | Dialysis |
Format | Cassette, Accessory |
Product Line | Slide-A-Lyzer™ |
Product Code | Brand | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||||
10770995
|
Thermo Scientific™
66430 |
For 0.5 to 3 mL Cassettes | Cassette Float Buoy |
£53.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10197393
|
Thermo Scientific™
66432 |
For 12 mL Cassettes | Cassette Float Buoy |
£43.69
Pack of 8 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10733495
|
Thermo Scientific™
66490 |
5 mL | Cassette Syringe |
£41.73
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10187493
|
Thermo Scientific™
66493 |
20 mL | Cassette Syringe |
£41.79
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10570904
|
Thermo Scientific™
66494 |
1 mL | Cassette Syringe |
£42.34
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Slide-A-Lyzer Dialysis Cassette Syringes are designed for efficient loading of samples into Slide-A-Lyzer cassettes and removal of sample after dialysis. Cassette Float Buoy accessories are for use with Slide-A-Lyzer Dialysis Cassettes used to remove low molecular weight contaminants and perform buffer exchange, desalting, and concentration.
Minimize sample loss into and out of Slide-A-Lyzer Dialysis Cassettes using Slide-A-Lyzer Dialysis Cassette Syringes for 1-mL to 20-mL sample volumes. Slide-A-Lyzer Cassette Float Buoys are designed for floating 0.5- to 12-mL cassettes in dialysate.Features of Slide-A-Lyzer Cassette Accessories include:
• Syringes are appropriately sized for different sample volumes
• Buoys keep stir bar from tearing dialysis membrane
• Matched to Slide-A-Lyzer Cassette for a secure fit and minimization of sample loss
• Chemically inert
Specifications
Store at room temperature. | |
Dialysis | |
Cassette, Accessory | |
Slide-A-Lyzer™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.