missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 2K MWCO
Remove salts and small contaminants to maximize sample purity and recovery with Slide-A-Lyzer 2K MWCO dialysis cassettes.
£111.00 - £143.00
Specifications
Content And Storage | Store at room temperature |
---|---|
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 2.0 kDa |
Product Code | Brand | Sample Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sample Volume (Metric) | Price | Quantity & Availability | |||||
10762325
|
Thermo Scientific™
66205 |
0.5 mL |
£111.00
Pack of 10 |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10371294
|
Thermo Scientific™
66203 |
3 mL |
£111.00
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10127483
|
Thermo Scientific™
66212 |
12 mL |
£143.00
Pack of 8 |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10465325
|
Thermo Scientific™
66230 |
30 mL |
£113.00
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Slide-A-Lyzer Dialysis Cassettes (2K MWCO) facilitate the simple and effective removal of salts and small contaminants from proteins and other macromolecules greater than 2,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange and complete sample recovery.
Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers. No knots, clips, or caps are needed to seal the units and prevent leaking and sample loss during the dialysis procedure.Features of Slide-A-Lyzer Dialysis Cassettes:
• Color-coded—blue frame specifies cassettes with the 2,000-MW pore-size membrane
• Four cassette sizes—select the cassette that best suits your sample volume (0.5 to 30 mL)
• Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
• High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
Membrane specifications:
• Composition: regenerated cellulose synthesized by the viscose method
• Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
• Hydration time required before use: 2 minutes for normal volumes or 4 minutes for minimal volumes
• Glycerol content: none
• Sulfur content: 0.169%
• Heavy metals content: trace
Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL
Specifications
Store at room temperature | |
Cellulose Membrane, Dialysis | |
2.0 kDa |
Buffer Exchange, Protein | |
Cassette | |
Slide-A-Lyzer™ |