missing translation for 'onlineSavingsMsg'
Få mere at vide

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 3.5K MWCO

Produktkode 10751565 Shop alle Thermo Scientific produkter
missing translation for 'orderingAttributeHoverText'
Sample Volume (Metric):
0.5 mL
3 mL
12 mL
30 mL
Quantity:
1 Kit (10 Cassettes)
1 Kit (8 Cassettes)
10 Cassettes
6 Cassettes
8 Cassettes
Förpackningsstorlek
Each
Pack of 10
Pack of 6
Pack of 8
Denne vare kan ikke returneres. Se returpolitik

Produktkode 10751565

missing translation for 'mfr': Thermo Scientific™ 66332

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

Remove salts and contaminants, and maximize sample recovery using this semi-permeable membrane with a 3,500-Dalton protein MW cut-off (MWCO) cassette that is easier to use than traditional dialysis tubing.

Slide-A-Lyzer Dialysis Cassettes (3.5K MWCO) facilitate simple and effective removal of salts and small contaminants from proteins and other macromolecules larger than 3,500 Daltons. They can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.

Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample loss during the dialysis procedure. Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume from any one of the four corner injection ports
Four cassette sizes—select the cassette that best suits your sample volume (0.5 to 30 mL)
Color-coded—pink cassette frame specifies cassettes with the 3,500 MW pore size membrane

Membrane specifications:
Composition: regenerated cellulose synthesized by the viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: none
Sulfur content: 0.1–0.15%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL

TRUSTED_SUSTAINABILITY

Tekniske data

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 3.5 kDa
Product Line Slide-A-Lyzer
Quantity 1 Kit (10 Cassettes)
Sample Volume (Metric) 3 mL
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.