missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 7K MWCO

Remove salts and contaminants and maximize sample recovery using this semi-permeable membrane with a 7,000-Dalton protein MW cut-off (7K MWCO).

Brand:  Thermo Scientific™ 66707

Code : PN

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10608854

  • £175.00 / Pack of 8
Estimated Shipment: 15-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Slide-A-Lyzer Dialysis Cassettes (7K MWCO) facilitate the simple and effective removal of salts and small contaminants from proteins or other macromolecules greater than 7,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 12 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange, and more complete sample recovery.

Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
Fast dialysis—flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
Four cassette sizes—select the cassette that best suits your sample volume
Color-coded—green cassette frame specifies cassettes with the 7,000 MW pore size membrane

Membrane specifications:
Composition: regenerated cellulose synthesized by the viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: 13%
Sulfur content: 0.1 to 0.15%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL

Specifications

Specifications

Dialysis Cassette Kit
Buffer Exchange, Protein
Cassette
Slide-A-Lyzer™
12 mL
Store at room temperature
Cellulose Membrane, Dialysis
7.0 kDa
1 Kit (8 Cassettes)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Slide-A-Lyzer Dialysis Cassette Technology is protected by U.S. Patent 5,503,741.